Läs mer
BI605906, MedChemExpress
Produktkod. 16455488
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16455488 10 mg 10mg
16411667 50 mg -
16408207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16455488 Leverantör Medchem Express Leverantörsnummer HY13019
MedChemExpress BI605906 is a novel IKKβ inhibitor with an IC50 value of 380 nM when assayed at 0.1 mM ATP.
BI605906 is a novel IKKβ inhibitor with an IC50 value of 380 nM when assayed at 0.1 mM ATP.
Specifikationer
Kemiskt namn eller material BI605906
CAS 960293-88-3
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C17H22F2N4O3S2
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 100 mg/mL (231.21 mM)
LEDER O=C(C1=C(N)C2=C(C(F)(F)CC)C=C(N3CCC(S(=O)(C)=O)CC3)N=C2S1)N
Molekylvikt (g/mol) 432.51
Formel vikt 432.51
Procent renhet 95.03%
Kvalitet Research
Fysisk form Solid