missing translation for 'onlineSavingsMsg'
Learn More

Bibenzyl, 99%, Thermo Scientific Chemicals

Catalog Number 15159163
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
103-29-7
C14H14
182.27
MFCD00004796
QWUWMCYKGHVNAV-UHFFFAOYSA-N
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
7647
CHEBI:34047
2-phenylethylbenzene
C(CC1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
103-29-7
C14H14
182.27
MFCD00004796
QWUWMCYKGHVNAV-UHFFFAOYSA-N
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
7647
CHEBI:34047
2-phenylethylbenzene
C(CC1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color White to Yellow
Density 1.0100g/mL
Boiling Point 284.0°C
Flash Point 129°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2C6H5
Quantity 500 g
Beilstein 05,598
Merck Index 15,1202
Specific Gravity 1.01
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in liquid ammonia, moderately soluble in alcohol, freely soluble in carbon disulfide, ether,, chloroform and amyl acetate, soluble in liquid sulfur dioxide
Formula Weight 182.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Bibenzyl, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'