Läs mer
BIBU 1361 Dihydrochloride, TRC
Produktkod. 30342700
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30342700 1 mg
1 options
Produktkod. 30342700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3770081MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material BIBU 1361 Dihydrochloride
CAS 1643609-75-9
Molekylformel C22H27ClFN7 • 2(HCl)
Kvantitet 1 mg
Synonym N-(3-Chloro-4-fluorophenyl)-6-[4-[(diethylamino)methyl]-1-piperidinyl]-pyrimido[5,4-d]pyrimidin-4-amine Dihydrochloride
InChI formel InChI=1S/C22H27ClFN7.2ClH/c1-3-30(4-2)13-15-7-9-31(10-8-15)22-25-12-19-20(29-22)21(27-14-26-19)28-16-5-6-18(24)17(23)11-16;;/h5-6,11-12,14-15H,3-4,7-10,13H2,1-2H3,(H,26,27,28);2*1H
LEDER CCN(CC1CCN(C2=NC=C3N=CN=C(C3=N2)NC4=CC(Cl)=C(C=C4)F)CC1)CC.[2HCl]
IUPAC-namn N-(3-chloro-4-fluorophenyl)-6-[4-(diethylaminomethyl)piperidin-1-yl]pyrimido[5,4-d]pyrimidin-4-amine;dihydrochloride
Molekylvikt (g/mol) 443.9523646