Läs mer
Bicyclomycin benzoate, MedChemExpress
Produktkod. 16499816
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16499816 50 mg -
16483278 10 mg -
16444536 100 mg 100mg
16412706 25 mg -
16406007 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16499816 Leverantör Medchem Express Leverantörsnummer HY101128
MedChemExpress Bicyclomycin benzoate is an antibiotic exhibiting activity against a broad spectrum of Gram-negative bacteria and against the Gram-positive bacterium.
Bicyclomycin benzoate is an antibiotic exhibiting activity against a broad spectrum of Gram-negative bacteria and against the Gram-positive bacterium.
Specifikationer
Kemiskt namn eller material Bicyclomycin benzoate
Fysisk form Solid
CAS 37134-40-0
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H22N2O8
För användning med (applikation) COVID-19-immunoregulation
Synonym FR2054
Löslighetsinformation DMSO : 100 mg/mL (246.07 mM; Need ultrasonic)
LEDER O[C@H]([C@](NC1=O)(OCCC2=C)C(N[C@]12O)=O)[C@@](C)(O)COC(C3=CC=CC=C3)=O
Molekylvikt (g/mol) 406.39
Formel vikt 406.39
Procent renhet 98.0%