missing translation for 'onlineSavingsMsg'
Learn More

Biebrich Scarlet sodium salt, Thermo Scientific Chemicals

Catalog Number p-7043119
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
4196-99-0
C22H14N4Na2O7S2
556.47
MFCD00003891
YCKMEQRBEVVZQF-VKZXTWAVSA-L
C.I. 26905; Acid Red 66
16219040
disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O
This item is not returnable. View return policy
4196-99-0
C22H14N4Na2O7S2
556.47
MFCD00003891
YCKMEQRBEVVZQF-VKZXTWAVSA-L
C.I. 26905; Acid Red 66
16219040
disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 188°C
Beilstein 3883900
Formula Weight 556.49
Color Brown to Red
Quantity 25 g
Flash Point 300°C (572°F)
Physical Form Powder
Chemical Name or Material Biebrich Scarlet sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'