Läs mer
Bifendate, TRC
Produktkod. 30344417
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30344417 1 g
1 options
Produktkod. 30344417 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3828901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bifendate
CAS 73536-69-3
Molekylformel C20 H18 O10
Kvantitet 1 g
Synonym 7,7'-Dimethoxy-[4,4'-Bi-1,3-benzodioxole]-5,5'-dicarboxylic Acid 5,5'-Dimethyl Ester, DDB, DDB, Dimethyl 4,4'-Dimethoxy-5,6,5',6'-di(methylenedioxy)biphenyl-2,2'-dicarboxylate, Dimethyl dicarboxylate Biphenyl, Diphenyl Dimethyl Bicarboxylate, α-DDB
InChI formel InChI=1S/C20H18O10/c1-23-11-5-9(19(21)25-3)13(17-15(11)27-7-29-17)14-10(20(22)26-4)6-12(24-2)16-18(14)30-8-28-16/h5-6H,7-8H2,1-4H3
LEDER O=C(C1=C(C2=C3OCOC3=C(C=C2C(OC)=O)OC)C4=C(OCO4)C(OC)=C1)OC
IUPAC-namn methyl 7-methoxy-4-(7-methoxy-5-methoxycarbonyl-1,3-benzodioxol-4-yl)-1,3-benzodioxole-5-carboxylate
Molekylvikt (g/mol) 418.35
Formel vikt 418.09