Läs mer
Bifonazole EP Impurity D (as Chloride), Mikromol™
Produktkod. 30052716
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052716 25 mg
30050999 100 mg
30463152 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30052716 Leverantör Mikromol Leverantörsnummer MM0318.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bifonazole EP Impurity D (as Chloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride
CAS 66600-13-3
Analyt- eller komponentnamn 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride
Frakt skick Room Temperature
Molekylformel C41 H33 N2 . Cl
Certifikat och efterlevnad ISO 9001
Synonym Bifonazole Imp. D (EP), 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride, Bifonazole Imp. D (EP) as Chloride, Bifonazole Impurity D as Chloride
InChI formel InChI=1S/C41H33N2.ClH/c1-5-13-32(14-6-1)34-21-25-38(26-22-34)40(36-17-9-3-10-18-36)42-29-30-43(31-42)41(37-19-11-4-12-20-37)39-27-23-35(24-28-39)33-15-7-2-8-16-33;/h1-31,40-41H;1H/q+1;/p-1
LEDER [Cl-].c1ccc(cc1)C(c2ccc(cc2)c3ccccc3)[n+]4ccn(c4)C(c5ccccc5)c6ccc(cc6)c7ccccc7
IUPAC-namn 1,3-bis[phenyl-(4-phenylphenyl)methyl]imidazol-1-ium;chloride
Molekylvikt (g/mol) 589.17
Kvantitet 25 mg
Formel vikt 588.2332 g/mol