Läs mer
Tocris Bioscience™ BigLEN (mouse)
GPR171 agonist
Brand: Tocris Bioscience™ 6304/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BigLEN (mouse) is a GPR171 agonist. ProSAAS-derived neuropeptide. Regulates food intake in mice. Inhibits the release of glutamate onto parvocellular neurons of the paraventricular nucleus in a process dependent upon activation of postsynaptic G proteins.Specifikationer
| 501036-69-7 | |
| Store at -20°C | |
| 1756.03 | |
| 1 mg |