missing translation for 'onlineSavingsMsg'
Läs mer

BIN2 Antibody, Biotin, Novus Biologicals™

Produktkod. 18029736 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
Förpackningsstorlek:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18029736 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18029736 Leverantör Novus Biologicals Leverantörsnummer NBP297015B

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

BIN2 Polyclonal specifically detects BIN2 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen BIN2
Användningsområden Western Blot, ELISA
Klassificering Polyclonal
Konjugera Biotin
Utspädning Western Blot, ELISA
Formulering PBS
Gene Alias BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2
Värd art Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BIN2 (Q9UBW5-1; Met1-Asn244).
Reningsmetod Affinity purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Cell Cycle and Replication
Primär eller sekundär Primary
Gen-ID (Entrez) 51411
Målarter Human
Innehåll och lagring Store at 4°C in the dark.
Form Purified
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.