Läs mer
Binimetinib, MedChemExpress
Produktkod. 16486666
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
200 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16486666 100 mg
16472477 50 mg
16436368 10 mg
16481166 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486666 Leverantör Medchem Express Leverantörsnummer HY15202
MedChemExpress Binimetinib (MEK162) is an oral and selective MEK1/2 inhibitor. Binimetinib (MEK162) inhibits MEK with an IC50 of 12 nM.
Binimetinib (MEK162) is an oral and selective MEK1/2 inhibitor. Binimetinib (MEK162) inhibits MEK with an IC50 of 12 nM.
Specifikationer
Kemiskt namn eller material Binimetinib
CAS 606143-89-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C17H15BrF2N4O3
Kvantitet 100 mg
Synonym MEK162 ARRY-162 ARRY-438162
Löslighetsinformation DMSO : 50 mg/mL (113.32 mM; Need ultrasonic)
LEDER BrC1=CC=C(C(F)=C1)NC2=C(C3=C(C=C2C(NOCCO)=O)N(C=N3)C)F
Molekylvikt (g/mol) 441.23
Formel vikt 441.23
Procent renhet 98.05%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease