Learn More
Hirschmann™ BIO-SILICO Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Hirschmann™ SILICOSEN Silicone Stoppers are specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.
Brand:  Hirschmann™ 8905917
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10362553
  • 710.00 SEK / Pack of 10
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Coarse pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Specifications
Orange
13-16 mm
10/Pk.
15 mm
Silicone Stopper
For The Preparation And Sterilization Of Culture Media
Silicone