Learn More
Ansell BioClean-D™ Non-sterile Cleanroom Coverall with Hood and Integrated Boots
Code produit 30058505
Modifier l'affichage
Click to view available options
Size:
X-Small
Conditionnement:
Pack of 20
Code produit Size Conditionnement
30058505 X-Small Pack of 20
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30058505 Fournisseur Ansell Code fournisseur BDFCXS
Beräknad leverans: 24-06-2027
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Cleanroom Coverall made of CleanTough™ fabric for lightweight, low-linting body protection with front zip and protective flap to provide contaminant and liquid splash barrier.
  • Reduced contamination risks: The BioClean-D Coverall with Hood and Integrated Boots BDFC is low-linting and ISO cleanroom-compliant thanks to its lightweight CleanTough™ composition, ensuring contamination risks are kept to a minimum
  • Optimized protection: This non-sterile disposable coverall features a front zip with a protective flap, which guards against liquid splashes and chemicals, while thumbs loops and an elasticated hood, back, cuffs and ankles offer a secure fit
  • Enhanced features: This antistatic coverall is also equipped with integrated boots and slip-resistant soles, for added workplace safety in laboratory environments
  • Reduced contamination risks: The BioClean-D Coverall with Hood and Integrated Boots BDFC is low-linting and ISO cleanroom-compliant thanks to its lightweight CleanTough™ composition, ensuring contamination risks are kept to a minimum
  • Optimized protection: This non-sterile disposable coverall features a front zip with a protective flap, which guards against liquid splashes and chemicals, while thumbs loops and an elasticated hood, back, cuffs and ankles offer a secure fit
  • Enhanced features: This antistatic coverall is also equipped with integrated boots and slip-resistant soles, for added workplace safety in laboratory environments
TRUSTED_SUSTAINABILITY
Spécification
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Flashspun Polyethylene
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static, Chemicals, Liquid, Particle
Collar Type Hooded
Certifications/Compliance (Europe) ASTM F739, CE 0598, EN ISO 13688:2013, EN 1149-5:2008, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009
Latex Free No
No. of Pockets None
For Use With (Application) Blending, compounding solids or liquids, Crime Scene Investigation (CSI), Forensics, Handling of Powders, Aseptic Filling and Labelling, Cleanroom Cleaning and Preparing, Medical Device Manufacturing, Biotechnologies Blending Operations, Handling Active Material, Maintaining Equipment and Instruments
Cleanroom Class Class 10 (ISO 4)
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.