missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ BioClean-D™ Sterile Drop-Down Cleanroom Coveralls with Hood

Catalog Number 15478320
Click to view available options
Size: X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

BioClean-D™ Sterile Drop-Down Cleanroom Coveralls with Hood include internal colored tabs to indicate safe touch points, strategically placed removable tabs, and a removable zip tab.

  • ISO Class 4 and EU GMP Grade A compatible
  • Antistatic, low linting durable CleanTough™ fabric
  • Quick release press stud tabs for touch-free donning
  • Elasticated back, hood, cuffs and ankles
  • Zip front
  • ASTM D6978 05 Tested
  • HELMKE DRUM Tested
  • Low-Particulate
  • Sterility Assurance Level 10−6
Product Type Cleanroom coverall
Size X-Large
Material CleanTough™ fabric
Color White
Disposable Yes
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Bound, Stitched
Sterility Sterile
Back Style Elastic
Certifications/Compliance ISO 9001:2015, (EU)2016/425, ASTM F739-12, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2, ISO 11137-1:2006
Description Quick release tabs, removable zip tab
Size (English) Chest 42 to 45 in.
Size (Metric) Chest 108 to 116 cm
For Use With (Application) Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities, Manufacturing, Industrial
Height (English) 6 ft. 0 in. to 6 ft. 2 in.
Height (Metric) 164 to 170 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'