Learn More
Nitritex™ BioClean™ Steele™ Cleanroom Shoes
Features antistatic soles, insoles in shoes for operator comfort.
Brand:  Nitritex™ BSTC-03
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15486154
  • 535.00 SEK / 1 pair
Estimated Shipment
31-01-2018
Add to basket
Description
Description
  • Slip-resistant
  • Antistatic soles and insoles
  • Steel toecap
  • Water; fuel-oil resistant
  • 200 joules impact resistance
  • Water penetration, absorption resistant
  • Energy absorbing seat region
  • Leather uppers, double density PU/PU antistatic injection molded soles
Quality standards:
  • Manufactured in a facility holding ISO 9001:2008
Certifications
EN 20345:2011
Videos