missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
BIOPHARMAGREENER_CHOICE

Catalog Number p-9045035
Click to view available options
Packaging:
20/Pk., 80/Cs.
5/Case
50/Cs
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Non-sterile; Lid: With Lid

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY
    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Array 16 x 24
    Packaging 5/Case
    Color Black, Clear Bottom
    Product Type Standard
    Material Polystyrene
    No. of Wells 384
    Sterility Non-sterile
    Surface Treatment Cell Culture
    Well Shape Round, Flat-bottom
    For Use With (Application) Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    Quantity 5/Case
    Source Poly-D-Lysine
    Well Color Clear
    Show More Show Less
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'