Läs mer
BioGEE, TRC
Produktkod. 30136294
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136294 100 mg
17907364 25 mg
17947754 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136294 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB397818100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 502635-76-9
Molekylformel C24 H39 N5 O8 S2
Molekylvikt (g/mol) 589.725
Synonym BioGEE
IUPAC-namn ethyl (2S)-2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-5-[[(2R)-1-[(2-ethoxy-2-oxoethyl)amino]-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoate
LEDER CCOC(=O)CNC(=O)[C@H](CS)NC(=O)CC[C@H](NC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C(=O)OCC
Specifikationer
Kemiskt namn eller material N-[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]-L-gamma-glutamyl-L-cysteinyl-glycine Diethyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C24H39N5O8S2/c1-3-36-20(32)11-25-22(33)15(12-38)27-19(31)10-9-14(23(34)37-4-2)26-18(30)8-6-5-7-17-21-16(13-39-17)28-24(35)29-21/h14-17,21,38H,3-13H2,1-2H3,(H,25,33)(H,26,30)(H,27,31)(H2,28,29,35)/t14-,15-,16-,17-,21-/m0/s1
Formel vikt 589.224