Läs mer
Biopterin, TRC
Produktkod. 30134309
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134309 25 mg
17971155 5 mg
17953685 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB38902025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22150-76-1
Molekylformel C9 H11 N5 O3
Molekylvikt (g/mol) 237.22
Synonym 4(1H)-Pteridinone, 2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-, 4(1H)-Pteridinone, 2-amino-6-(1,2-dihydroxypropyl)-, [S-(R*,S*)]-, 4(3H)-Pteridinone, 2-amino-6-(L-erythro-1,2-dihydroxypropyl)- (7CI,8CI), 2-Amino-6-[(1R,2S)-1,2-dihydroxypropyl]-4(1H)-pteridinone, (-)-Biopterin, 6-Biopterin, Biopterin, L-Biopterin, L-erythro-Biopterin, NSC 339699, Pterin H B2
IUPAC-namn 2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-3H-pteridin-4-one
LEDER C[C@H](O)[C@H](O)c1cnc2N=C(N)NC(=O)c2n1
Specifikationer
Kemiskt namn eller material 6-Biopterin
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C9H11N5O3/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7/h2-3,6,15-16H,1H3,(H3,10,11,13,14,17)/t3-,6-/m0/s1
Formel vikt 237.0862
Procent renhet >95%