Learn More
Unity Lab Services™ Biosafety Cabinet Ki Test 3rd Party
Product Code. 30286553
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
Product Code. Duration Unit Size
30286553 1 Day Each
1 options
Product Code. 30286553 Supplier Unity Lab Services™ Supplier No. BSCKIMVS
 Restricted Item
Beräknad leverans: 29-12-2026
KI disc testing for biosafety cabinets, manufactured by third parties such as Labconco and Erlab. This will provide reassurance of cabinet performance and validation of containment efficiency.
  • KI disc testing for biosafety cabinets
  • Manufactured by third parties such as Labconco and Erlab.
  • This will provide reassurance of cabinet performance and validation of containment efficiency.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinets
Duration 1 Day
Product Type Installation, Connection, and Commissioning Service