missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Synergy H4 Hybrid Microplate Reader
Combines two powerful detection systems, monochromator-based and filter-based, in one compact unit
POA
Specifications
Accuracy | ±1% at 2 OD, typical |
---|---|
Dynamic Range | 0 to 99,999 (fluorescence, luminescence); 0 to 4.0 O.D. (absorbance) |
Emission Wavelength Range | 300 to 700 nm (800 nm with red extended option) |
Excitation Wavelength Range | 200 to 700 nm |
Hertz | 50/60 Hz |
Description
Enjoy complete flexibility and assay choice for current as well as future demands. Advanced multi-detection system is capable of performing an unlimited number of microplate-based assays.
- Hybrid Technology: Synergy H4 combines the sensitivity of a filter-based system with the convenience of monochromator-based optics to provide the broadest range of applications available on the market today.
- Detection modes: Fluorescence Intensity, Time-Resolved Fluorescence, Fluorescence Polarization, AlphaScreen/AlphaLISA, Luminescence, UV-Visible absorbance, FRET, TR-FRET, BRET, well area scanning and spectral scanning.
- Modular and upgradeable architecture: Read modes are available as individual modules for cost-effectiveness and peace of mind.
- Quadruple mono system and variable bandpass selection: Synergy H4 optics incorporates two double-grating monochromators. This quadruple design provides the best in spectral scanning performance and flexibility.
- Deep blocking filters and dichroic mirrors: Synergy H4's filter/dichroic combination provides the best possible performance in fluorescence, time resolved fluorescence and fluorescence polarization applications.
- Compatible with Take3 Plate with 2μL microspots: Enables low volume 260 nm nucleic acid quantification
Synergy H4 Monochromator System
- Uses two double-grating monochromators with variable bandpass selection
- Highest stray light reflection, continuous wavelength selection, spectral scanning
- Combines high performance with convenience and flexibility
- Delivers more energy to the sample and provides high signal-to-noise rations
- Faster read speed, more sensitivity, more precise control over optical parameters
Certifications
CE Marked, ETL Listed
Specifications
±1% at 2 OD, typical | |
300 to 700 nm (800 nm with red extended option) | |
50/60 Hz | |
≥1% from 0 to 3.0 O.D. (typical) | |
100/240 V |
0 to 99,999 (fluorescence, luminescence); 0 to 4.0 O.D. (absorbance) | |
200 to 700 nm | |
Tungsten Halogen Lamp, Xenon Flash Lamp | |
2.5pg/mL SF; typical |