Läs mer
(+)-Biotin Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (+)-Biotin Methyl Ester |
| CAS | 608-16-2 |
| Molekylformel | C11 H18 N2 O3 S |
| Kvantitet | 10 mg |
| Synonym | D-(+)-Biotin Methyl Ester, Methyl Biotinate, d-Biotin Methyl Ester, Biotin Methyl Ester, [3aS-(3aα,4β,6aα)]-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic Acid Methyl Ester, (3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic Acid Methyl Ester |
| InChI formel | InChI=1S/C11H18N2O3S/c1-16-9(14)5-3-2-4-8-10-7(6-17-8)12-11(15)13-10/h7-8,10H,2-6H2,1H3,(H2,12,13,15)/t7?,8-,10?/m0/s1 |
| LEDER | COC(=O)CCCC[C@@H]1SCC2NC(=O)NC12 |
| IUPAC-namn | methyl 5-[(4S)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate |
| Molekylvikt (g/mol) | 258.34 |
| Formel vikt | 258.1038 |