missing translation for 'onlineSavingsMsg'
Learn More

(+)-Biotin Methyl Ester, TRC

Product Code. 30330393
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30330393 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330393 Supplier Toronto Research Chemicals Supplier No. TRCB390480100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (+)-Biotin Methyl Ester
CAS 608-16-2
Molekylformel C11 H18 N2 O3 S
Kvantitet 100 mg
Synonym D-(+)-Biotin Methyl Ester, Methyl Biotinate, d-Biotin Methyl Ester, Biotin Methyl Ester, [3aS-(3aα,4β,6aα)]-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic Acid Methyl Ester, (3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic Acid Methyl Ester
InChI formel InChI=1S/C11H18N2O3S/c1-16-9(14)5-3-2-4-8-10-7(6-17-8)12-11(15)13-10/h7-8,10H,2-6H2,1H3,(H2,12,13,15)/t7?,8-,10?/m0/s1
LEDER COC(=O)CCCC[C@@H]1SCC2NC(=O)NC12
IUPAC-namn methyl 5-[(4S)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
Molekylvikt (g/mol) 258.34
Formel vikt 258.1038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.