Läs mer
Tocris Bioscience™ Biotin-NAD+
Beskrivning
Biotin-NAD+ is a provides a convenient non-isotopic alternative to radiolabeled NAD+ for determination of IC50 values for candidate PARP inhibitors and studies requiring this substrate. Biotinylated-NAD+ allows an indirect measure of PARP activity when biotin incorporation is detected using a conjugated-streptavidin detection system. Acts as a substrate for ADP-ribosylation. Can be used to label and purify biotinyl-ADP ribosylated proteins.
This product is a replacement for R&D Systems product 4670-500-01 .
131 μg is supplied as 500 μl of a 0.25 mM solution in water.
Specifikationer
Specifikationer
| Kemiskt namn eller material | β-Nicotinamide-N6-[2-[[6-[biotinyl]amino]hexyl]amino]-2-oxoethyl]adenine dinucleotide |
| CAS | 146385-37-7 |
| Kvantitet | 131 μg |
| Molekylformel | C39H57N11O17P2S |