Learn More
Invitrogen™ Biotin-Ubiquitin
Biotin-Ubiquitin
3155.00 SEK - 14140.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11545016
|
Invitrogen™
PV4379 |
10μg |
3155.00 SEK
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11555016
|
Invitrogen™
PV4380 |
100μg |
14140.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).
For Research Use Only. Not for use in diagnostic procedures.