missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34771PE
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Biotin (Vitamin B7 or Vitamin H) Monoclonal specifically detects Biotin (Vitamin B7 or Vitamin H) in All Species samples. It is validated for Flow Cytometry.
Specifikationer
| Biotin (Vitamin B7 or Vitamin H) | |
| Monoclonal | |
| PE | |
| 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
| Biotinylated sheep immunoglobulin | |
| Protein A or G purified | |
| RUO | |
| It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This monoclonal antibody is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SPM375 | |
| Flow Cytometry | |
| Mouse | |
| 244 kDa | |
| 0.1 mL | |
| Primary | |
| All species | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering