Läs mer
Molecular Probes™ Biotin-X Cadaverine (5-(((N-(Biotinoyl)Amino)Hexanoyl)Amino)Pentylamine, Trifluoroacetic Acid Salt)
Beskrivning
The primary aliphatic amine of biotin-X cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
Specifikationer
Specifikationer
| Produkttyp | Biotin-X Cadaverine |
| Kvantitet | 10 mg |
| Innehåll och lagring | Store at room temperature. |
| Frakt skick | Room Temperature |
| Kemisk reaktivitet | Carboxylic Acid, Ketone, Aldehyde |
| Reaktiv del | Amine, Cadaverine |
| Etikett eller färgämne | Biotin-X |
| Etiketttyp | Biotin & Other Haptens |