Läs mer
Biotinyl Cystamine Hydrochloride, TRC
Produktkod. 30339263
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30339263 100 mg
1 options
Produktkod. 30339263 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB395800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Biotinyl Cystamine Hydrochloride
CAS 1346597-28-1
Molekylformel C14H27ClN4O2S3
Kvantitet 100 mg
Synonym (3aS,4S,6aR)-N-[2-[(2-Aminoethyl)dithio]ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide Hydrochloride
InChI formel InChI=1S/C14H26N4O2S3.ClH/c15-5-7-22-23-8-6-16-12(19)4-2-1-3-11-13-10(9-21-11)17-14(20)18-13;/h10-11,13H,1-9,15H2,(H,16,19)(H2,17,18,20);1H/t10?,11-,13?;/m0./s1
LEDER Cl.NCCSSCCNC(=O)CCCC[C@@H]1SCC2NC(=O)NC12
IUPAC-namn N-[2-(2-aminoethyldisulfanyl)ethyl]-5-[(4S)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanamide;hydrochloride
Molekylvikt (g/mol) 415.04
Formel vikt 414.0985