missing translation for 'onlineSavingsMsg'
Läs mer

[Biotinylamidoethyl]-dithiomethylenemalonic Acid Bis(2-aminoethanoic Acid), TRC

Produktkod. 30328900
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30328900 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30328900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB39421050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material [Biotinylamidoethyl]-dithiomethylenemalonic Acid Bis(2-aminoethanoic Acid)
CAS 1217718-11-0
Molekylformel C22H34N6O9S3
Kvantitet 50 mg
InChI formel InChI=1S/C22H34N6O9S3/c29-14(4-2-1-3-13-18-12(11-38-13)26-22(37)28-18)23-6-8-40-39-7-5-15(30)27-19(20(35)24-9-16(31)32)21(36)25-10-17(33)34/h12-13,18-19H,1-11H2,(H,23,29)(H,24,35)(H,25,36)(H,27,30)(H,31,32)(H,33,34)(H2,26,28,37)/t12-,13-,18-/m0/s1
LEDER OC(=O)CNC(=O)C(NC(=O)CCSSCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C(=O)NCC(=O)O
IUPAC-namn 2-[[2-[3-[2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethyldisulfanyl]propanoylamino]-3-(carboxymethylamino)-3-oxopropanoyl]amino]acetic acid
Molekylvikt (g/mol) 622.74
Formel vikt 622.1549
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.