Learn More
ACROBiosystems Biotinylated Human Her2 / ErbB2 Protein, His, Avitag™
Biotinylated Protein
Brand: ACROBiosystems HE2-H82E2-200UG
Additional Details : Weight : 0.00001kg
Description
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 73.0 kDa. The protein migrates as 85-95 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_004439.2 | |
73.0 kDa | |
200 µg | |
Thr 23 - Thr 652 | |
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1 | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>90% as determined by SDS-PAGE | |
Biotinylated Human Her2, His,Avitag (HE2-H82E2) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). | |
-20°C | |
Her2 | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |