missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human NKG2C&CD94 Protein, His, Avitag

Brand:  ACROBiosystems NC4-H82E3-25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17216340

  • 4673.08 SEK / 25µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. NKG2C/CD159c is used as a receptor for NK cells and some cytotoxic T cells to recognize MHC class I HLA-E molecules. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.3 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

34.3 kDa
Biotinylated Human NKG2C&CD94;, His,Avitag (NC4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) (Accession # P26717-1 (NKG2C) & Q13241-1 (CD94)).
-20°C
NKG2C & CD94
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
25 μg
Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94)
NKG2C & CD94
Precision Avi
His Tag & Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers