Läs mer
Biperiden hydrochloride, Tocris Bioscience™
Produktkod. 18702714
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18702714 10 mg
18782754 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18702714 Leverantör Tocris Bioscience Leverantörsnummer 2853/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Muscarinic receptor antagonist; some selectivity for M1
Muscarinic receptor antagonist that displays some selectivity for the M1 subtype (Ki values are 0.48, 2.4, 3.9, 6.3 and 6.3 nM for M1, M4, M3, M2 and M5 receptors respectively). Alleviates extrapyramidal symptoms associated with antipsychotic drugs. Displays antiparkinsonian activity.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1235-82-1
Molekylformel C21H30ClNO
Molekylvikt (g/mol) 347.927
InChI-nyckel RDNLAULGBSQZMP-LIJKWCKLSA-N
Synonym 1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
PubChem CID 129318274
IUPAC-namn (1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
LEDER C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl
Specifikationer
Kemiskt namn eller material Biperiden hydrochloride
Kvantitet 10 mg
Formel vikt 347.92
Procent renhet >99%