missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-3-carboxaldehyde, 95%, Thermo Scientific™

Catalog Number p-7033353
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1204-60-0
C13H10O
182.222
MFCD01740432
KFKSIUOALVIACE-UHFFFAOYSA-N
1,1'-biphenyl-3-carbaldehyde, biphenyl-3-carbaldehyde, 3-formylbiphenyl, ccris 3164, biphenyl-3-carboxaldehyde, 3-difenilaldeide italian, 1,1'-biphenyl-3-carboxaldehyde, 3-biphenyl carboxaldehyde
121053
3-phenylbenzaldehyde
C1=CC=C(C=C1)C2=CC(=CC=C2)C=O
This item is not returnable. View return policy
1204-60-0
C13H10O
182.222
MFCD01740432
KFKSIUOALVIACE-UHFFFAOYSA-N
1,1'-biphenyl-3-carbaldehyde, biphenyl-3-carbaldehyde, 3-formylbiphenyl, ccris 3164, biphenyl-3-carboxaldehyde, 3-difenilaldeide italian, 1,1'-biphenyl-3-carboxaldehyde, 3-biphenyl carboxaldehyde
121053
3-phenylbenzaldehyde
C1=CC=C(C=C1)C2=CC(=CC=C2)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material Biphenyl-3-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'