missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BIRC5 (Human) Recombinant Protein
Human BIRC5 full-length ORF ( AAH08718.1, 1 a.a. - 142 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000332-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGAPTLPPAWQPFLKDHRISTFKNWPFLEGCACTPERMAEAGFIHCPTENEPDLAQCFFCFKELEGWEPDDDPIEEHKKHSSGCAFLSVKKQFEELTLGEFLKLDRERAKNKIAKETNNKKKEFEETAKKVRRAIEQLAAMD
Specifications
AAH08718.1 | |
baculoviral IAP repeat-containing 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIRC5 | |
GST |
332 | |
Wheat germ expression system | |
10 μg | |
API4, EPR-1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |