Läs mer
bis(1,2,2,6,6-Pentamethyl-4-piperidyl) Sebacate, TRC
Produktkod. 30135179
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135179 5 g
30133053 10 g
30007388 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2755085G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41556-26-7
Molekylformel C30H56N2O4
Molekylvikt (g/mol) 508.77
Synonym ADK Stab LA 72, Adekastab LA 72, Antioxidant 292, BASF 292, Bis(1,2,2,6,6-pentamethyl-4-piperdinyl)-sebacate, Bis(1,2,2,6,6-pentamethyl-4-piperidinyl) sebacate;Bis(1,2,2,6,6-pentamethyl-4-piperidyl) 1,8-Octanedicarboxylate, Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Decanedioate, Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Sebacate, Bis(1-methyl-2,2,6,6-tetramethyl-4-piperidinyl) Sebacate, Bis(N-methyl-2,2,6,6-tetramethyl-4-piperidinyl) Sebacate, Chiguard 353, Chisorb 292;HALPS 292, HALS 4, HS 508, JCK-W-US, LA 72, LA 77ME, LS 292, LS 508, LS 765;Light Stabilizer 292, Light Stabilizer 765, Lowilite 76, Sanol 292, Sanol LS 292, Sanol LS 508, Sanol LS 765, Songlight 2920LQ, T 292, TIN 292, TN 765, Tinuvin 292, Tinuvin 765, Tinuvin 766, UV 55-07051, UV 765;
IUPAC-namn bis(1,2,2,6,6-pentamethylpiperidin-4-yl) decanedioate
LEDER CN1C(C)(C)CC(CC1(C)C)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)N(C)C(C)(C)C2
Specifikationer
Kemiskt namn eller material bis(1,2,2,6,6-Pentamethyl-4-piperidyl) Sebacate
Kvantitet 5 g
InChI formel InChI=1S/C30H56N2O4/c1-27(2)19-23(20-28(3,4)31(27)9)35-25(33)17-15-13-11-12-14-16-18-26(34)36-24-21-29(5,6)32(10)30(7,8)22-24/h23-24H,11-22H2,1-10H3
Formel vikt 508.424