Läs mer
Bis(1,3-dichloro-2-propyl) Phosphate, TRC
Produktkod. 30339456
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30339456 5 mg
1 options
Produktkod. 30339456 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4190955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(1,3-dichloro-2-propyl) Phosphate
CAS 72236-72-7
Anteckningar om renhetsgrad HPLC
Molekylformel C6 H11 Cl4 O4 P
Kvantitet 5 mg
Synonym 2-Propanol, 1,3-dichloro-, hydrogen phosphate (9CI), Bis(1,3-dichloro-2-propyl) phosphate, 1,3-Dichloro-2-propanol Hydrogen Phosphate, 2-Propanol, 1,3-dichloro-, hydrogen phosphate (9CI)
InChI formel InChI=1S/C6H11Cl4O4P/c7-1-5(2-8)13-15(11,12)14-6(3-9)4-10/h5-6H,1-4H2,(H,11,12)
LEDER OP(=O)(OC(CCl)CCl)OC(CCl)CCl
IUPAC-namn bis(1,3-dichloropropan-2-yl) hydrogen phosphate
Molekylvikt (g/mol) 319.93
Formel vikt 317.9149