Läs mer
Bis(2,3-dichloropropyl) Ether, TRC
Product Code. 30331644
Change view
Click to view available options
Kvantitet:
100 mg
Product Code. Kvantitet
30331644 100 mg
1 options
Product Code. 30331644 Supplier Toronto Research Chemicals Supplier No. TRCB426220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Bis(2,3-dichloropropyl) Ether
CAS 7774-68-7
Molekylformel C6 H10 Cl4 O
Kvantitet 100 mg
Synonym Bis(2,3-dichloropropyl) Ether, Propane, 1,2-dichloro-3-(2,3-dichloropropoxy)-, Ether, bis(2,3-dichloropropyl) (8CI), Propane, 1,1'-oxybis[2,3-dichloro- (9CI), 1,2-Dichloro-3-(2,3-dichloropropoxy)propane, Bis(2,3-Dichloropropyl) ether
InChI formel InChI=1S/C6H10Cl4O/c7-1-5(9)3-11-4-6(10)2-8/h5-6H,1-4H2
LEDER ClCC(Cl)COCC(Cl)CCl
IUPAC-namn 1,2-dichloro-3-(2,3-dichloropropoxy)propane
Molekylvikt (g/mol) 239.96
Formel vikt 237.9486