Läs mer
Bis(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate Tetrahydrochloride, TRC
Produktkod. 30344468
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30344468 100 mg
1 options
Produktkod. 30344468 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB222760100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate Tetrahydrochloride
CAS 2227064-51-7
Molekylformel C54H61N6O6.4HCl
Kvantitet 100 mg
Synonym Bis[2-(4-benzhydrylpiperazin-1-yl)ethyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate Tetrahydrochloride, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3,5-bis[2-[4-(diphenylmethyl)-1-piperazinyl]ethyl] ester, hydrochloride (1:4), Bis[2-[4-(diphenylmethyl)-1-piperazinyl]ethyl] 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-pyridine-3,5-dicarboxylate Tetrahydrochloride
InChI formel InChI=1S/C53H58N6O6.4ClH/c1-39-47(52(60)64-36-34-55-26-30-57(31-27-55)50(41-16-7-3-8-17-41)42-18-9-4-10-19-42)49(45-24-15-25-46(38-45)59(62)63)48(40(2)54-39)53(61)65-37-35-56-28-32-58(33-29-56)51(43-20-11-5-12-21-43)44-22-13-6-14-23-44;;;;/h3-25,38,49-51,54H,26-37H2,1-2H3;4*1H
LEDER O=C(C1=C(C)N([H])C(C)=C(C1C2=CC(N(=O)=O)=CC=C2)C(OCCN3CCN(C(C4=CC=CC=C4)C5=CC=CC=C5)CC3)=O)OCCN6CCN(C(C7=CC=CC=C7)C8=CC=CC=C8)CC6.[H]Cl.[H]Cl.[H]Cl.[H]Cl
IUPAC-namn bis[2-(4-benzhydrylpiperazin-1-yl)ethyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;tetrahydrochloride
Molekylvikt (g/mol) 1020.91