Läs mer
Bis(2,4-dimethylphenyl) Disulfide, TRC
Produktkod. 30387019
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30387019 10 mg
30383551 50 mg
30375978 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30387019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48866010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13616-83-6
Molekylformel C16H18S2
Molekylvikt (g/mol) 274.44
Synonym 2,4-Xylyl disulfide (6CI,7CI,8CI), 1,2-Bis(2,4-dimethylphenyl)disulfane
IUPAC-namn 1-[(2,4-dimethylphenyl)disulfanyl]-2,4-dimethylbenzene
LEDER Cc1ccc(SSc2ccc(C)cc2C)c(C)c1
Specifikationer
Kemiskt namn eller material Bis(2,4-dimethylphenyl) Disulfide
Kvantitet 10 mg
InChI formel InChI=1S/C16H18S2/c1-11-5-7-15(13(3)9-11)17-18-16-8-6-12(2)10-14(16)4/h5-10H,1-4H3
Formel vikt 274.085