Läs mer
Bis[2-(4-hydroxyphenylthio)ethoxy]methane, TRC
Produktkod. 17984145
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17984145 500 mg
17971875 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17984145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB447430500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93589-69-6
Molekylformel C17H20O4S2
Molekylvikt (g/mol) 352.47
Synonym 4,4'-[Methylenebis(oxy-2,1-ethanediylthio)]bis[phenol], 1,7-Bis(4-hydroxyphenylthio)-3,5-dioxaheptane, Bis[2-(4-hydroxyphenylthio)ethoxy]methane
IUPAC-namn 4-[2-[2-(4-hydroxyphenyl)sulfanylethoxymethoxy]ethylsulfanyl]phenol
LEDER Oc1ccc(SCCOCOCCSc2ccc(O)cc2)cc1
Specifikationer
Kemiskt namn eller material Bis[2-(4-hydroxyphenylthio)ethoxy]methane
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C17H20O4S2/c18-14-1-5-16(6-2-14)22-11-9-20-13-21-10-12-23-17-7-3-15(19)4-8-17/h1-8,18-19H,9-13H2
Procent renhet >95%