Läs mer
Bis[2,5-Dimethoxy-4-(p-nitrophenylazo)benzenediazonium] Chlorozincate, TRC
Produktkod. 30376184
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30376184 1 g
1 options
Produktkod. 30376184 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD6407111G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis[2,5-Dimethoxy-4-(p-nitrophenylazo)benzenediazonium] Chlorozincate
CAS 64071-86-9
Molekylformel 2[C14 H12 N5 O4] . [Cl4 Zn]
Kvantitet 1 g
Synonym Fast Black K Salt Hemi(zinc chloride) Salt, Tetrachloro-bis[2,5-dimethoxy-4-[(4-nitrophenyl)azo]benzenediazonium] Zincate, 2,5-Dimethoxy-4-(4-nitrophenyl)diazenylbenzenediazonium Dichlorozinc, 2,5-Dimethoxy-4-[(4-nitrophenyl)azo]-tetrachlorozincate(2-) Benzenediazonium (2:1)
InChI formel InChI=1S/2C14H12N5O4.4ClH.Zn/c2*1-22-13-8-12(14(23-2)7-11(13)16-15)18-17-9-3-5-10(6-4-9)19(20)21;;;;;/h2*3-8H,1-2H3;4*1H;/q2*+1;;;;;+2/p-4
LEDER [Cl-][Zn+2]([Cl-])([Cl-])[Cl-].COc1cc([N+]#N)c(OC)cc1N=Nc2ccc(cc2)N(=O)=O.COc3cc([N+]#N)c(OC)cc3N=Nc4ccc(cc4)N(=O)=O
Molekylvikt (g/mol) 835.77
Formel vikt 831.9824