Läs mer
Bis(2-aminoethylthio)methane Dihydrochloride, TRC
Produktkod. 30334526
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30334526 5 g
1 options
Produktkod. 30334526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4022005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(2-aminoethylthio)methane Dihydrochloride
CAS 22965-82-8
Molekylformel C5H14N2S2.2(HCl)
Kvantitet 5 g
Synonym 2,2'-[Methylenebis(thio)]bisethanamine Dihydrochloride
InChI formel InChI=1S/C5H14N2S2.2ClH/c6-1-3-8-5-9-4-2-7;;/h1-7H2;2*1H
LEDER NCCSCSCCN.Cl.Cl
IUPAC-namn 2,2'-(methylenebis(sulfanediyl))bis(ethan-1-amine) Dihydrogen Chloride
Molekylvikt (g/mol) 239.22
Formel vikt 238.02