missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Bis(2-butoxyethyl) Phthalate (~85%), TRC
Specifications
Specifications
| Kemiskt namn eller material | Phthalic acid,bis-butoxyethyl ester |
| CAS | 117-83-9 |
| Molekylformel | C20 H30 O6 |
| Kvantitet | 1 g |
| Synonym | 1,2-Benzenedicarboxylic acid, 1,2-bis(2-butoxyethyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-butoxyethyl) ester (9CI), Phthalic acid, bis(2-butoxyethyl) ester (6CI,7CI,8CI), Ethanol, 2-butoxy-, phthalate (2:1) (8CI), Bis(2-butoxyethyl) phthalate, Bis(2-n-butoxyethyl) phthalate, Butyl glycol phthalate, DBMP, Di(2-butoxyethyl) phthalate, Di(ethylene glycol butyl ether) phthalate, Kesscoflex BCP, Kronisol, NSC 4840, Palatinol K, Plasthall 200DBEP, Plasthall DBEP, Sankonol 0882-0, Sansocizer C 1100, β-Butoxyethyl phthalate |
| InChI formel | InChI=1S/C20H30O6/c1-3-5-11-23-13-15-25-19(21)17-9-7-8-10-18(17)20(22)26-16-14-24-12-6-4-2/h7-10H,3-6,11-16H2,1-2H3 |
| LEDER | CCCCOCCOC(=O)c1ccccc1C(=O)OCCOCCCC |
| IUPAC-namn | bis(2-butoxyethyl) benzene-1,2-dicarboxylate |
| Molekylvikt (g/mol) | 366.45 |
| Formel vikt | 366.2042 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?