Läs mer
Bis(2-butoxyethyl) Phthalate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid,bis-butoxyethyl ester |
| CAS | 117-83-9 |
| Molekylformel | C20 H30 O6 |
| Kvantitet | 100 mg |
| Synonym | 1,2-Benzenedicarboxylic acid, 1,2-bis(2-butoxyethyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-butoxyethyl) ester (9CI), Phthalic acid, bis(2-butoxyethyl) ester (6CI,7CI,8CI), Ethanol, 2-butoxy-, phthalate (2:1) (8CI), Bis(2-butoxyethyl) phthalate, Bis(2-n-butoxyethyl) phthalate, Butyl glycol phthalate, DBMP, Di(2-butoxyethyl) phthalate, Di(ethylene glycol butyl ether) phthalate, Kesscoflex BCP, Kronisol, NSC 4840, Palatinol K, Plasthall 200DBEP, Plasthall DBEP, Sankonol 0882-0, Sansocizer C 1100, β-Butoxyethyl phthalate |
| InChI formel | InChI=1S/C20H30O6/c1-3-5-11-23-13-15-25-19(21)17-9-7-8-10-18(17)20(22)26-16-14-24-12-6-4-2/h7-10H,3-6,11-16H2,1-2H3 |
| LEDER | CCCCOCCOC(=O)c1ccccc1C(=O)OCCOCCCC |
| IUPAC-namn | bis(2-butoxyethyl) benzene-1,2-dicarboxylate |
| Molekylvikt (g/mol) | 366.45 |
| Formel vikt | 366.2042 |