Läs mer
Bis(2-chloroethyl)aminophosphoric Dichloride, TRC
Produktkod. 30342693
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30342693 250 mg
1 options
Produktkod. 30342693 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB418810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bis(2-chloroethyl)aminophosphoric Dichloride
CAS 127-88-8
Molekylformel C4 H8 Cl4 N O P
Kvantitet 250 mg
Synonym Phosphoramidic dichloride, N,N-bis(2-chloroethyl)- (ACI), N,N-Bis(2-chloroethyl)phosphoramidic dichloride (ACI), Phosphoramidic dichloride, bis(2-chloroethyl)- (7CI, 8CI, 9CI), (N,N-Di-2-chloroethyl) phosphoramidic dichloride, 2-Chloro-N-(2-chloroethyl)-N-dichlorophosphorylethanamine, Bis(2-chloroethyl)aminophosphoric dichloride, Bis(2-chloroethyl)aminophosphoryl dichloride, Bis(2-chloroethyl)phosphoramidic dichloride, N,N-Bis(2-chloroethyl)phosphamide dichloride, NSC 64119
InChI formel InChI=1S/C4H8Cl4NOP/c5-1-3-9(4-2-6)11(7,8)10/h1-4H2
LEDER ClCCN(CCCl)P(=O)(Cl)Cl
IUPAC-namn 2-chloro-N-(2-chloroethyl)-N-dichlorophosphorylethanamine
Molekylvikt (g/mol) 258.9
Formel vikt 256.9098