Läs mer
Bis(2-ethoxyethyl) Phthalate (>90%), TRC
Produktkod. 30343361
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30343361 5 mg
1 options
Produktkod. 30343361 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3997855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-2-ethoxyethyl ester
CAS 605-54-9
Molekylformel C16 H22 O6
Kvantitet 5 mg
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-ethoxyethyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-ethoxyethyl) ester (9CI), Phthalic acid, bis(2-ethoxyethyl) ester (6CI,7CI,8CI), Ethanol, 2-ethoxy-, phthalate (2:1) (8CI), Bis(2-ethoxyethyl) phthalate, Bis(ethoxyethyl) phthalate, Di-2-ethoxyethyl phthalate, Diethoxyethyl phthalate, NSC 4825, Phthalic acid bis(2-ethoxyethyl) ester, BEEP
InChI formel InChI=1S/C16H22O6/c1-3-19-9-11-21-15(17)13-7-5-6-8-14(13)16(18)22-12-10-20-4-2/h5-8H,3-4,9-12H2,1-2H3
LEDER CCOCCOC(=O)c1ccccc1C(=O)OCCOCC
IUPAC-namn bis(2-ethoxyethyl) benzene-1,2-dicarboxylate
Molekylvikt (g/mol) 310.34
Formel vikt 310.1416