Läs mer
Bis(2-Ethylbutyl) Phthalate, TRC
Produktkod. 30011394
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011394 100 mg
30004938 1 g
30005832 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678780100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7299-89-0
Molekylformel C20H30O4
Molekylvikt (g/mol) 334.45
Synonym Phthalic Acid Bis(2-ethylbutyl) Ester, Di(2-ethylbutyl) Phthalate, NSC 15317
LEDER O=C(OCC(CC)CC)C1=CC=CC=C1C(OCC(CC)CC)=O
Specifikationer
Kemiskt namn eller material Bis(2-Ethylbutyl) Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C20H30O4/c1-5-15(6-2)13-23-19(21)17-11-9-10-12-18(17)20(22)24-14-16(7-3)8-4/h9-12,15-16H,5-8,13-14H2,1-4H3
Formel vikt 334.2144
Procent renhet >95%