Läs mer
Bis(2-ethylhexyl) Phthalate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, bis-2-ethylhexyl ester |
| CAS | 117-81-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C24 H38 O4 |
| Kvantitet | 10 g |
| Synonym | 1,2-Benzenedicarboxylic acid, bis(2-ethylhexyl) ester (9CI), Phthalic acid, bis(2-ethylhexyl) ester (6CI,8CI), 2-Ethylhexyl phthalate, 3315AF2, Bis(2-ethylhexyl) 1,2-benzenedicarboxylate, Bis(2-ethylhexyl) o-phthalate, Bis(2-ethylhexyl) phthalate, Bisoflex 81, Bisoflex DOP, Codan Set L 86P, Compound 889, Corflex 400, DEHP, DOF, DOF (Russian plasticizer), DOP, DOPT, Di(2-ethylhexyl) phthalate, Di(isooctyl) phthalate, Diacizer DOP, Dioctyl phthalate, Diplast O, ESBO-D 82, Ergoplast FDO, Ergoplast FDO-S, Etalon, Etalon (plasticizer), Ethylhexyl phthalate, Eviplast 80, Eviplast 81, Fleximel, Flexol DOD, Flexol DOP, Garbeflex DOP-D 40, Good-rite GP 264, Hatco DOP, JSSD-DOP, Jayflex DOP, Kodaflex DEHP, Kodaflex DOP, Monocizer DOP, NSC 17069, Octoil, Octyl phthalate, Palatinol AH, Palatinol AH-L, Palatinol DOP, Phthalic acid di(2-ethylhexyl) ester, Phthalic acid dioctyl ester, Pittsburgh PX 138, Plasthall DOP, R 8000, Reomol D 79P, Sansocizer DOP, Sansocizer R 8000, Scandinol SC 1000, Sconamoll DOP, Sicol 150, Staflex DOP, Truflex DOP, Vestinol AH, Vinycizer 80, Vinycizer 80K, Witcizer 312, ZS, ZS (plasticizer) |
| InChI formel | InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3 |
| LEDER | CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC |
| IUPAC-namn | bis(2-ethylhexyl) benzene-1,2-dicarboxylate |
| Molekylvikt (g/mol) | 390.56 |
| Visa mer |