Läs mer
Bis(2-hydroxyethyl) Terephthalate, TRC
Produktkod. 30133350
Change view
Kvantitet:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133350 100 g
30009560 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133350 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB443860100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 959-26-2
Molekylformel C12 H14 O6
Molekylvikt (g/mol) 254.236
Synonym 1,4-Benzenedicarboxylic acid, bis(2-hydroxyethyl) ester (9CI), Ethylene glycol, terephthalate (2:1) (8CI), Terephthalic acid, bis(2-hydroxyethyl) ester (6CI, 8CI), BHET, BHET(J), BHET-N, Bis(2-hydroxyethyl) 1,4-benzenedicarboxylate, Bis(2-hydroxyethyl) te
IUPAC-namn bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
LEDER OCCOC(=O)c1ccc(cc1)C(=O)OCCO
Specifikationer
Kemiskt namn eller material Terephthalic Acid, Bis-2-hydroxyethyl Ester
Kvantitet 100 g
InChI formel InChI=1S/C12H14O6/c13-5-7-17-11(15)9-1-2-10(4-3-9)12(16)18-8-6-14/h1-4,13-14H,5-8H2
Formel vikt 254.079