missing translation for 'onlineSavingsMsg'
Learn More

Bis-(2-methanethiosulfonatoethyl)methylamine, TRC

Product Code. 30337477
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30337477 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337477 Supplier Toronto Research Chemicals Supplier No. TRCB4859105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Bis-(2-methanethiosulfonatoethyl)methylamine
CAS 16216-82-3
Molekylformel C7 H17 N O4 S4
Kvantitet 5 mg
InChI formel InChI=1S/C7H17NO4S4/c1-8(4-6-13-15(2,9)10)5-7-14-16(3,11)12/h4-7H2,1-3H3
LEDER CN(CCSS(=O)(=O)C)CCSS(=O)(=O)C
IUPAC-namn N-methyl-2-methylsulfonylsulfanyl-N-(2-methylsulfonylsulfanylethyl)ethanamine
Molekylvikt (g/mol) 307.47
Formel vikt 307.004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.