Läs mer
Bis(2-methoxyethyl) Phthalate-3,4,5,6-d4, TRC
Produktkod. 30332120
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30332120 1 mg
1 options
Produktkod. 30332120 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4875111MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-methylglycol ester D4
CAS 1398065-54-7
Molekylformel C14 2H4 H14 O6
Kvantitet 1 mg
Synonym Phthalic Acid, Bis-methylglycol Ester D4, Phthalic Acid, Bis-methylglycol Ester D4 (phenyl D4)
InChI formel InChI=1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3/i3D,4D,5D,6D
LEDER [2H]c1c([2H])c([2H])c(C(=O)OCCOC)c(C(=O)OCCOC)c1[2H]
IUPAC-namn bis(2-methoxyethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvikt (g/mol) 286.31
Formel vikt 286.1354