Läs mer
Bis(2-propylheptyl) Phthalate, TRC
Produktkod. 30134661
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134661 1 g
17912695 100 mg
17951685 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5235001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53306-54-0
Molekylformel C28 H46 O4
Molekylvikt (g/mol) 446.66
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-propylheptyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-propylheptyl) ester (9CI), Phthalic acid, bis(2-propylheptyl) ester (6CI), Bis(2-propylheptyl) phthalate, DPHP, Di-2-propylheptyl phthalate, Dipropylheptyl phthalate, Hexamoll DPHP, NSC 17071, Palatinol 10P, Phthalic acid, bis-2-propylheptyl ester
IUPAC-namn bis(2-propylheptyl) benzene-1,2-dicarboxylate
LEDER CCCCCC(CCC)COC(=O)c1ccccc1C(=O)OCC(CCC)CCCCC
Specifikationer
Kemiskt namn eller material Bis(2-propylheptyl) Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C28H46O4/c1-5-9-11-17-23(15-7-3)21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-24(16-8-4)18-12-10-6-2/h13-14,19-20,23-24H,5-12,15-18,21-22H2,1-4H3
Formel vikt 446.3396
Procent renhet >95%